Learn More
Merck Millicell μ-Angiogenesis Activation Assay Kit
Product Code. 11790363 Shop All Merck Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11790363 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11790363 Supplier Merck Supplier No. MMA130
Estimated Shipment: 02-09-2026
Specifications
Detection Method Fluorescence
Includes Millicell™ μ-Angiogenesis Slides Five tissue culture treated slides. Each slide contains 15 wells, Calcein-AM One 50 μg vial, PMA One 1mg vial. ITS Media Supplement, 100X One 0.5mL vial, Thrombin Solution One 0.75mL vial, Fibrinogen Solution
Quantity 1 Kit
Packaging 1 Kit
For Use With (Application) Angiogenesis Assays
Host Species Vertebrates, Human, Rat
Product Type Millicell μ-Angiogenesis Activation Assay Kit