Learn More
Sperian™ Honeywell™ Millenia™ Sport Safety Glasses
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Unit Size:
Each
Description
- Universal fit
- Excellent optical properties
- Lightweight value-based frame gives superior side and brow protection
- Comes standard with a Flexicord™ retainer
- Available with either anti-fog or anti-scratch lens coating
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Product Type | Millennia Sport |
| Frame Color | Black |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |