Learn More
Miele Miele DOS K85 / 1 flex dosing module short suction lance (217mm)
Brand: Miele 11498920
590.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Miele DOS K85 / 1 flex dosing module short suction lance (217mm)