missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas Midi Horizontal Gel Electrophoresis Unit
Description
- Plastic casting gates
- Colour-coded combs
- Quick-release lid to disconnect tank and lid simultaneously
- UV-transparent acrylic, for direct visualisation of gel within tank
- Compact design providing optimal control over voltage gradient and run-time
- Small size. allows safe and easy transportation
Specifications
Specifications
| Type | Gel Unit |
| Gel Dimensions | 150 x 128 mm |
| Includes | 1 x gel casting tray, 2 x 1.0mm 16 sample combs, buffer recirculation ports, power supply connectors and loading strips |
| Sample Size | 112 |
| Volume (Metric) Buffer | 900 mL |
| Length (Metric) Gel | 150 mm |
| Width (Metric) Gel | 128 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?