Learn More
Camlab™ Microtube Biohazard Label
Specifications
Specifications
| Product Type | Microtube Biohazard Label |
| No. per Pack | 250/Pk. |
| Size (Metric) | 11 x 14 mm |
| Temperature Range (Metric) | 5 to 80°C |
| Diameter (Metric) | 9.5 mm |