Learn More
Kinematica™ Microtron™ MB 800 Laboratory Mixer
Product Code. 11979058 Shop All Kinematica Products
Click to view available options
Voltage:
230 V
Unit Size:
Each
Product Code. 11979058
Brand: Kinematica™ 36010003
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A laboratory mixer well-suited for intense mixing, dispersing, homogenizing, emulsifying and mincing operations for cuttable materials, making samples easy to work with for analysis and quality control.
  • Smooth rotational speed adjustment with integrated on/off switch
  • Load compensated rotational speed adjustment
  • Switching of mixing attachment and protection cover without tools
  • Operation only possible with security cover and lid in place
  • Variety of sample container designs
  • Vibration dampening quick coupling
  • Very quiet motor with integrated ventilation cooling
  • Integrated load, heat and blockage protection
  • Production of very fine particle sizes in both liquid and dry mediums - precise analyses require homogenous samples
  • Efficient sample preparation through high rotational speeds
  • Flexibility with various sample volumes with different mixing attachments
  • Durable, robust, long-lasting design
Specifications
Product Type Laboratory Mixer
Speed 1000 to 17,000 rpm
Voltage 230 V
Hertz 60 Hz
Wattage 800 W
Volume Range (Metric) 2 to 4 L
Height (Metric) 245 mm
For Use With (Application) for Mixing, Dispersing, Homogenizing, Emulsifying and Crushing of Foodstuffs
Model MB 800 with EU Power Cable
Noise Level 79 dBA at 17,000 rpm
Packaging Quantity 1 Ea.
Weight (Metric) 5.3 kg