Learn More
Deltalab™ Sterile Microtitre Plates
Product Code. p-8009132
Click to view available options
Well Shape:
Flat Bottom (F-well)
Unit Size:
Pack of 50
Product Code. 12168910
Brand: Deltalab™ 900011
Only null left
Add to basket
Edge
Add to basket
Use Deltalab™ Sterile Microtitre Plates for applications including diluting, serial sample preparation, protein precipitation and hemagglutination. The plates are used when optical reading is required.
  • Plate dimensions: 127.7 x 85.8 mm.
  • Conical bottom plates may be used for complement fixation
  • Polystyrene lids supplied separately
  • Plates can be covered using sealing mat or the adhesive sealing film
Specifications
Format Standard
Product Type “U” form
Sterility Sterile
Well Shape Flat Bottom (F-well)
No. per Pack 50
Quantity 50/Pk.