Learn More
Carl Zeiss™ AxioCams Microscopy Camera
Click to view available options
Description:
AxioCam 105 Color
AxioCam ERc 5s
Unit Size:
Each
Description
- CMOS microscope camera, 5 megapixels (2560 x 1920)
- Sensor size of 1/2.5 in. (diagonal 7.1 mm)
- Pixel size 2.2 x 2.2 μm
- Color camera
- Includes ZEN Lite starter software for image capture and making measurements
Specifications
Specifications
| Type | Microscopy Camera AxioCam |
| Description | AxioCam 105 Color |
| Includes | Axiocam 105 color, driver software 64-bit, USB 3.0 PCIe x1 interface, USB 3.0 connection cable (3 m), SATA power cable and Molex power cable sensor |