Learn More
Rogo Sampaic™ Microscopic Needle
Product Code. 11708234
Change view
Click to view available options
Length (Metric):
140 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11708234 140 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708234 Supplier Rogo Sampaic™ Supplier No. FMD005
  • ONLINE EXCLUSIVE PRICE
    14.60€
    12.55€ / Pack of 10
    Save 2.05€
    14% Off
Estimated Shipment: 18-11-2026
Specifications
Product Type Microscopic needle
Material Stainless Steel
Tip Style Curved Point
Length (Metric) 140 mm