Learn More
Carl Zeiss™ Microscope Stage Micrometers
Product Code. 11884953
Change view
Click to view available options
For Use With (Equipment):
Reflected light microscope, eyepiece graticule
Transmitted light microscope, eyepiece graticule
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11884953 Transmitted light microscope, eyepiece graticule Each
11866422 Reflected light microscope, eyepiece graticule Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11884953 Supplier Carl Zeiss™ Supplier No. 4740260000000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Includes
5 lines with 1mm intervals in y-direction, 100 lines with 1/100mm intervals in y-direction
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
  • For calibrating optical systems with eyepiece graticule patterns
  • 5 lines with 1mm intervals in y-direction
  • 100 lines with 1/100mm intervals in y-direction
Specifications
Type Stage Micrometer
For Use With (Equipment) Transmitted light microscope, eyepiece graticule
Includes Stage micrometer, 5 lines with 1mm intervals in y-direction