Learn More
DuraLab Microscope Micrometer Eyepiece WF10x/22 mm for DX Series
Product Code. 30280252
Click to view available options
Type:
Widefield Eyepiece
Unit Size:
Each
Product Code. 30280252
Brand: DuraLab DRL0061
Only null left
Add to basket
Add to basket
Micrometer eyepiece for DX Series microscopes, designed for precise specimen measurement and accurate scientific analysis.
  • Supports precise measurement of microscopic specimens, helping users quantify size and scale accurately
  • Assists students in developing measurement skills by observing and comparing structures at a microscopic level
  • Facilitates calibration of eyepiece graticules, ensuring reliable and consistent results across scientific applications
Specifications
Type Widefield Eyepiece
Compatible with DRL0045 - DuraLab DX-820 Pro Infinity Binocular Microscope, DRL0046 - DuraLab DX-830 Pro Infinity Trinocular Microscope