Learn More
DuraLab Microscope Micrometer Eyepiece WF10x/22 mm for DX Series
Click to view available options
Type:
Widefield Eyepiece
Unit Size:
Each
Description
- Supports precise measurement of microscopic specimens, helping users quantify size and scale accurately
- Assists students in developing measurement skills by observing and comparing structures at a microscopic level
- Facilitates calibration of eyepiece graticules, ensuring reliable and consistent results across scientific applications
Specifications
Specifications
| Type | Widefield Eyepiece |
Compatible with DRL0045 - DuraLab DX-820 Pro Infinity Binocular Microscope, DRL0046 - DuraLab DX-830 Pro Infinity Trinocular Microscope