Learn More
Sartorius Microsart™ Media
Product Code. p-4700605 Shop All Sartorius Products
Click to view available options
Without Additives:
No Cholesterol
No Sucrose
Unit Size:
Pack of 100
Product Code. 15952510
Brand: Sartorius 1432247ACN
Enhance the safety of microbiological quality control in the pharmaceutical and biotech industries for microbial limits testing according to the USP and EP
  • Ready-to-use agar media dish (prefilled with different agar media types) with active lid
  • Sterile packaged and ready to use together with Microsart filter units
  • Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
  • Liftable interior lid provides easy access to select colonies after incubation for further analysis
TRUSTED_SUSTAINABILITY
Specifications
Classification Reduced Growth Factor, Very Low Protein (<90ng/mL)
Form Gel
Product Type Growth Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Gamma-irradiated
With Additives Essential and nonessential amino acids
Without Additives No Sucrose
For Use With (Application) Target microorganisms: total count, stressed organisms found in pure water systems
Formulation Solid Agar Medium