Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Description
Thermo Scientific™ Microplate Reader Verification Plates are designed to be convenient solutions for verifying and documenting the optical performance of compatible Thermo Scientific™ microplate readers. The portfolio includes options for photometric, fluorometric, and luminometric performance verification, helping laboratories maintain confidence in measurement quality and instrument consistency over time.
Available verification plates
- Multifunctional Verification Plate (N03394M2): Verifies photometric and fluorometric performance. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers. Supported tests depend on the instrument model and may include photometric accuracy, precision, wavelength accuracy (verifies simultaneously fluorometric excitation wavelength accuracy) and stray light. With fluorometry, both top and bottom sensitivity and fluorescence emission wavelength accuracy can be verified.
- Spectrophotometric Verification Plate (N03394): Verifies photometric accuracy, precision, linearity, wavelength accuracy, and stray light. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers.
- Multiskan Verification Plate, 8 wavelengths (24072800) or 16 wavelengths (24072805): Verifies photometric performance across 8 or 16 wavelengths. Compatible with Multiskan™ Ease (24072800) and Multiskan™ FC (24072800 or 24072805) readers.
- Lumiwell Luminometric Verification Plate (2806460): Provides an independent Relative Light Unit reference for luminometric performance verification. Compatible with Varioskan™ LUX and Varioskan™ ALF readers.
The verification plates include certified optical reference materials appropriate for the supported detection technology. Photometric reference values are traceable to standards from the National Physical Laboratory (NPL, UK) and/or the National Institute of Standards and Technology (NIST, USA), as specified for each product. A Certificate of Calibration is supplied to support verification documentation.
The products include ready-to-use Thermo Scientific™ SkanIt™ Software sessions loading results automatically to Microsoft™ Excel™ data analysis template files to simplify measurement, result evaluation, and documentation.
Key features
- Options for photometric, fluorometric, and luminometric verification
- Supports routine performance verification and documentation
- Certified reference values for reliable result evaluation
- Designed for compatible Thermo Scientific™ microplate readers
- Preconfigured SkanIt Software sessions and analysis files included
Includes:
- Verification plate
- Certificate of Calibration
- Verification plate adapter, where required
- USB drive with sessions, reference and data analysis template files
Protective carrying case
Specifications
Specifications
| Product Type | Multiskan Verification Plate (Abs), 16 Wavelengths |
| For Use With (Equipment) | Multiskan Ease, Multiskan FC |
| Plate Type | Verification Plate |
| Wavelength | 16 Wavelengths |
| Includes | NPL-traceable Multiskan Verification Plate, Calibration certificate, USB with SkanIt sessions and reference files, Carrying case |