Learn More
OHAUS™ Microplate Holder
Shop All Ohaus Products
Click to view available options
Holds:
1 Standard Microplate
4 Standard Microplates
Two standard microplates
Unit Size:
Each
Description
- For use with Ohaus Microplate Vortex Mixer
Specifications
Specifications
| Holds | 1 Standard Microplate |
| Product Type | Heavy-Duty Vortex |
| Material | Plasitc |
| Description | Single Microplate Holder |
| For Use With (Application) | Re-Suspending Liquids, Mixing Reagents, Buffers and Emulsions, ELISA Assays, Drug Extractions, Mixing Samples, PCR, Digestions, Cell Disruptions, Genotyping |
| For Use With (Equipment) | Microplate Vortex Mixer/Heavy Duty Vortex Mixer |
| Packaging Quantity | 1 Ea. |