Learn More
VITLAB™ Piston-Operated Micropipettes
Click to view available options
Volume (Metric):
100 to 1000 μL
1000 to 10,000 μL
500 to 5000 μL
Accuracy:
±30μL
±6μL
±60μL
Unit Size:
Each
Description
- Completely autoclavable at 121°C acc. to DIN EN 285
- Highly accurate with simple calibration for long-lasting reliability
- Large, central pipetting button provides a uniform and smooth movement of the piston
- Ergonomic eject button for rapid tip replacement
- 4-digit volume display with integrated zoom function and vertical arrangement of the numbers ensures optimal readability at all times
- Desired volume can be set by rotating the volume setting wheel with ease and precision
- Integrated calibration function allows adjustment without tools directly in the laboratory
- Corrosion-resistant piston and ejector ensure long product life
- Includes micropipette, silicone grease, sample bag with pipette tips, quality certificate and operating manual
Specifications
Specifications
| Accuracy | ±6μL |
| Autoclavable | Autoclavable |
| Type | Piston Operated Micropipette |
| Volume (Metric) | 100 to 1000 μL |
| Display | 4-digit |
| Includes | Micropipette, silicone grease, sample bag with pipette tips, quality certificate, operating manual |