Learn More
Micronova™ MegaTape™ Polyethylene Cleanroom Tape
Product Code. 13167326 Shop All Micronova Products
Click to view available options
Width (Metric):
25.4 mm
50.8 mm
Unit Size:
Each
Product Code. 13167326
Brand: Micronova™ MTP2WH
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A medium density, polyethylene tape with hi-tack/clean-peel acrylic adhesive, good conformability and excellent resistance to moisture, oils and weathering. Ideal for clean construction and maintenance projects.
The Micronova MegaTape™ Polyethylene Cleanroom Tape, with hi-tack acrylic adhesive, provides superior sealing abilities and removes cleanly with no adhesive residue. This tape is low outgassing, has good conformability, similar to vinyl tapes, and shows excellent resistance to UV, moisture, acids, alkalis, oils, abrasion and weathering.
Specifications
Autoclavable Not Autoclavable
Adhesive Type Acrylic
Color White
Material Polyethylene
Product Type Cleanroom Tape
Sterility Non-sterile
For Use With (Application) Ideal for Clean Construction and Maintenance Projects
Temperature Range (English) 60°F to 80°F
Temperature Range (Metric) 16°C to 27°C
Dimensions (LxW) 33 m x 50.8 mm (36 yd. x 2 in.)
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 2 in.
Width (Metric) 50.8 mm