Learn More
Micronova™ Hand Held Scraper
Product Code. 13127286 Shop All Micronova Products
Change view
Click to view available options
Length (English):
8 in.
9.25 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
13127286 8 in. Each
13177786 9.25 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13127286 Supplier Micronova™ Supplier No. HHSWH8
  • ONLINE EXCLUSIVE PRICE
    21.70€
    20.81€ / Pack of 1
    Save 0.89€
    4% Off
 Restricted Item
Estimated Shipment: 04-01-2027
A hand held, polypropylene scraper tool with an injection-molded, seamless design that prevents bacterial growth during residue and build-up removal in controlled environments.
The Micronova Hand Held Scraper features injection-molded, polypropylene construction, without seams or cracks that might harbor bacterial growth. The scraper is autoclavable and will not scratch metal surfaces.
  • Injection-molded, hand held scraper tool
  • For removing residue and build up in controlled environments
  • White polypropylene construction
  • No seams or cracks to harbor bacterial growth
  • Will not scratch metal surfaces
  • Available in 8 or 10-inch lengths
  • Autoclavable
Specifications
Product Type Scraper Tool
Color White
Length (English) 8 in.
Sterility Non-sterile
Length (Metric) 20.3 cm
Autoclavable Autoclavable
Width (Metric) 7.6 cm
Width (English) 3 in.
Resistant To Chemicals
Size 7.62 x 20.32 (3 x 8 in.)
Disposable Reusable
Quantity 1/Pk., 12/Cs.
Care Instructions Store at Ambient Temperature
For Use With (Application) Controlled Environment Residue Removal
Certifications/Compliance FDA Compliant
Color-coded No