Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
Product Code. 10746732
Click to view available options
Color:
Clear
Binding Property:
High Binding
Medium Binding
Unit Size:
Pack of 100
Product Code. 10746732
Brand: Greiner Bio-One 705071
Only null left
Add to basket
Add to basket
For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Individual wells can be pipetted without leaving residue
  • Flat bottom enables precise optical measurements
  • Suitable for transmission measurements
  • Specifica
Service Plan
No
Warranty
No
TRUSTED_SUSTAINABILITY
Specifications
Breakapart Single break
Binding Property High Binding
Format Strip Plate
Array 8 x 12
Packaging Inner bags of 10
Binding Type High Binding
Color Clear
Product Type 96-well Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon
Well Shape Curved Edges, Flat Bottom (C-well)
Dnase Free Yes
For Use With (Application) Colorimetric immunoassay
Quantity 100/Cs.
RNase Free Yes
Dimensions (L x W x H) 127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)