Learn More
Socorex™ Acura™ Self-refill 865 Microdispensing Pipette
Description
- Hand-held.
- Three-way valve system for automatic self-refilling.
- Durable and resistant to aggressive media.
- Excellent ergonomics.
- Ultra-soft plunger activation.
- Large volume display.
- Versatile feeding through bottle, tubing or syringe.
- Easy in-lab calibration.
- Individually serial numbered and Q.C. certified.
- Autoclavable.
Specifications
Specifications
| Accuracy | <±2.5% - <±1.0% |
| Autoclavable | Autoclavable |
| Number of Channels | 1 |
| Product Line | Acura |
| Type | Microdispenser |
| Volume (Metric) | 20 to 200 μL |
| Precision | <1.5% - <0.3% |
| Resistance | Aggressive media |
| Warranty | 36 Months |