missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauda LAUDA Microcool MC 600 Circulation chille 230 V, 50 Hz EU plug

Brand: Lauda L001048
3868.01 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24121" to get your promotional price.
LAUDA Microcool MC 600 Circulation chille 230 V, 50 Hz EU plug
This item is not returnable.
View return policy
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction