Learn More
Thermo Scientific™ MicroClick 24 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Codice prodotto. 15195283 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Capacity:
24 x 2 mL
Dimensione della confezione:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacity Dimensione della confezione
15195283 24 x 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15195283 Fornitore Thermo Scientific™ N. del fornitore 75005715
  • ONLINE EXCLUSIVE PRICE
    1760.00€
    1376.41€ / Pack of 1
    Save 383.59€
    21% Off
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Perform micro-volume applications such as nucleic acid preparation at up to 16,000rpm/ 24,372 x g with the Thermo Scientific™ MicroClick 24 x 2 microtube rotor.
Perform micro-volume applications such as nucleic acid preparation at up to 17,850rpm/ 30,279 x g with the Thermo Scientific™ MicroClick 24 x 2 microtube rotor.
With Auto-Lock rotor exchange, switch from swing-out to a microtube rotor in as little as 3 seconds with:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
Biocontainment rotor lid for glove-friendly, one-handed operation, certified by Public Health England, Porton Down, UK.
Ordering Alerts:
Replacement biocontainment lid (75005725) and replacement O-rings (75003405) are sold separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifica
Description MicroClick 24 x 2 Fixed Angle Microtube Rotor
Type Fixed Angle Rotor
Capacity 24 x 2 mL
Max. RCF 24,372 x g
Max. Speed 16,000 rpm
Radius (Metric) 85 mm
K Factor 505
Max. Tube Length 45 mm
Max. Tube Diameter 11 mm
For Use With (Equipment) Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle 45°
Biocontainment Yes
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.