Learn More
Ansell MICROCHEM™ 6000
Product Code. 17150955| Product Code. | Size | Style | unitSize |
|---|---|---|---|
| 17150955 | 3X-Large | O60A812E | Each |
| 17100915 | Large | O60T801B | Each |
| 17100925 | Small | O60T803E | Each |
| 17100945 | Large | O60T809E | Each |
| 17100955 | Small | O60A812E | Each |
| 17100965 | 2X-Large | O60A813E | Each |
| 17100975 | Large | O60A818E | Each |
| 17110915 | X-Large | O60T801B | Each |
| 17110925 | Medium | O60T803E | Each |
| 17110945 | X-Large | O60T809E | Each |
| 17110955 | Medium | O60A812E | Each |
| 17110965 | 3X-Large | O60A813E | Each |
| 17110975 | X-Large | O60A818E | Each |
| 17120915 | 2X-Large | O60T801B | Each |
| 17120925 | Large | O60T803E | Each |
| 17120935 | Small | O60T808E | Each |
| 17120945 | 2X-Large | O60T809E | Each |
| 17120955 | Large | O60A812E | Each |
| 17120975 | 2X-Large | O60A818E | Each |
| 17130915 | 3X-Large | O60T801B | Each |
| 17130925 | X-Large | O60T803E | Each |
| 17130935 | Medium | O60T808E | Each |
| 17130945 | 3X-Large | O60T809E | Each |
| 17130955 | X-Large | O60A812E | Each |
| 17130975 | 3X-Large | O60A818E | Each |
| 17140915 | Small | O60T802E | Each |
| 17140925 | 2X-Large | O60T803E | Each |
| 17140935 | Large | O60T808E | Each |
| 17140955 | 2X-Large | O60A812E | Each |
| 17140975 | Small | O60A819E | Each |
| 17150915 | Medium | O60T802E | Each |
| 17150925 | 3X-Large | O60T803E | Each |
| 17150935 | X-Large | O60T808E | Each |
| 17150975 | Medium | O60A819E | Each |
| 17160915 | Large | O60T802E | Each |
| 17160935 | 2X-Large | O60T808E | Each |
| 17160955 | Small | O60A813E | Each |
| 17160975 | Large | O60A819E | Each |
| 17170915 | X-Large | O60T802E | Each |
| 17170935 | 3X-Large | O60T808E | Each |
| 17170955 | Medium | O60A813E | Each |
| 17170975 | X-Large | O60A819E | Each |
| 17180905 | Small | O60T801B | Each |
| 17180915 | 2X-Large | O60T802E | Each |
| 17180935 | Small | O60T809E | Each |
| 17180955 | Large | O60A813E | Each |
| 17180965 | Small | O60A818E | Each |
| 17180975 | 2X-Large | O60A819E | Each |
| 17190905 | Medium | O60T801B | Each |
| 17190915 | 3X-Large | O60T802E | Each |
| 17190935 | Medium | O60T809E | Each |
| 17190955 | X-Large | O60A813E | Each |
| 17190965 | Medium | O60A818E | Each |
| 17190975 | 3X-Large | O60A819E | Each |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Description
- Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
- Innovative fabric technology, high performance barrier laminate with scrim reinforcement
- Lightweight and flexible
- Highly visible bright orange color for improved worker safety
- Ultrasonically welded and hot air taped seams
- DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
- The zip has an outer zip flap made of the same material as the suit
- Semi-rigid 3 layer GAG visor provides excellent optical clarity
Specifications
Specifications
| Color | Orange |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning |
| Material | Multi-Layer Non-Woven Barrier Laminate Fabric |
| Size | 3X-Large |
| Seam Type | Welded and Taped |
| Style | O60A812E |
| Weight (Metric) | 230 gsm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.