Learn More
Cytiva Microcheck™ Beverage Monitor with Metricel™ Black Membrane
Product Code. 17134401
Click to view available options
Pore Size:
0.8 μm
Unit Size:
Pack of 50
Product Code. 17134401
Brand: Cytiva 4764
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
  • Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish
  • Not approved for use in medical clinical surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Pore Size 0.8 μm
Quantity 50/Pk.
Sterility Sterile
Product Type Gridded Filtering Funnel
Material (Membrane) Metricel Mixed Cellulose Ester