Learn More
Metro™ Microban Rack
Product Code. 11745769
Change view
Click to view available options
Dimensions (L x W):
1220 x 455 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11745769 1220 x 455 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11745769 Supplier Metro™ Supplier No. 5Q357G2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Rack
Diameter (Metric) 455 mm
Depth (Metric) 455 mm
Dimensions (L x W) 1220 x 455 mm
For Use With (Application) Inhibits The Growth Of Mold And Bacteria
Height (Metric) 189.5 cm
Length (Metric) 122 cm
Load Bearing Capacity (Metric) 270 kg
Width (Metric) 455 mm