Learn More
Biochrom Micro-Volume Cuvette
Click to view available options
For Use With (Equipment):
15 mm BH Spectorphotometers
8.5 mm BH Spectrophotometers
Pathlength:
0.125 mm
0.2 mm
0.5 mm
Unit Size:
Each
Description
- Exceptional measurement accuracy
- Wide measurement range - no more changing pathlengths
- Use only 1 μL of preicous sample
- Simple to use and clean
- Easy to check for sample impurities such as bubbles or dust
- Compatible with most standatd UV-Visible Sprectrophotometer
Specifications
Specifications
| Description | Micro-Volume Cuvette with 0.2 mm Pathlength for 15 mm Beam Height Spectrophotometers |
| Detection Limits | 1.2 ng/μL DNA |
| Dimensions (L x W x H) | 12.5 x 12.5 x 61.0 mm |
| Type | Microvolume Cuvette |
| Disposable | Reusable |
| For Use With (Application) | Life Science |
| For Use With (Equipment) | 15 mm BH Spectorphotometers |
| Pathlength | 0.2 mm |
| Sample Volume (Metric) | Minimum 0.8 μL |
| Shape | Rectangular |
For research use only.