Learn More
SI Analytics™ Micro-Ubbelohde Viscometers
Product Code. 10632441 Shop All SI Analytics Products
Click to view available options
Viscosity Range:
4 to 60 cSt
40 to 800 cSt
Unit Size:
Each
Product Code. 10632441
Brand: SI Analytics™ 285401025
Only null left
Add to basket
Add to basket
SI Analytics™ Micro-Ubbelohde Viscometers feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, minimum measurement substance and/or rinsing agent quantities, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • Due to their design, these viscometers are especially suitable for measurement of small liquid quantities and for particularly short running times
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • This constant is determined by multiplication of the constant K with the correction factor F
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ubbelohde Micro Viscometer
Viscosity Range 4 to 60 cSt
Calibrated Yes
Description 0.70 mm diameter
Approximate Constant 0.1
Certifications/Compliance DIN 51562, Part 2
Material Glass
Length (Metric) 290 mm
For Use With (Application) Manual measurements
Volume (Metric) 3 to 4 mL