Learn More
SI Analytics™ Micro Ubbelohde Viscometer
Product Code. 10090071 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
0.4 to 6 cSt
12 to 180 cSt
4 to 60 cSt
40 to 800 cSt
Approximate Constant:
0.01K
0.1K
0.3K
1K
Size:
0.4mm Diameter
0.70mm Diameter
0.95mm Diameter
1.26mm Diameter
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Approximate Constant Size Unit Size
10090071 0.4 to 6 cSt 0.01K 0.4mm Diameter Each
10537371 4 to 60 cSt 0.1K 0.70mm Diameter Each
10159990 12 to 180 cSt 0.3K 0.95mm Diameter Each
10121721 40 to 800 cSt 1K 1.26mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10090071 Supplier SI Analytics™ Supplier No. 285401103
In Stock
Determine the limit viscosity number of polymers. SI Analytics™ Micro Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, high or low temperature measurements, and minimum measurement substance or rinsing agent quantities
  • The recommended minimum flow through time is 200 seconds
  • Calibrated
  • With constant for automatic measurements
  • Filling quantity of 3 to 4mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 0.4 to 6 cSt
Calibrated Yes
Approximate Constant 0.01K
Certifications/Compliance DIN 55 350, Part 18 and DIN 51562, Part 2
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative, Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 0.4mm Diameter
Volume (Metric) 3 to 4 mL