missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Micro Tubes

Product Code. 30158425
Click to view available options
:
With Hinged Snap Cap
This item is not returnable. View return policy

Product Code. 30158425

Brand: Heinz Herenz 2000006

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polypropylene micro tubes 1.5 mL. With safe-lock cap, conical bottom, and molded graduation.

  • Micro tube
  • 1.5 mL
  • PP
  • With safe-lock cap
  • Conical bottom
  • Molded graduation
  • Basic-Line
  • 500 pieces per bag

Specifications

Autoclavable Not Autoclavable
Color Transparent
Material Polypropylene
Packaging Carton
Sterility Non-sterile
Graduated Yes
Closure Type Hinged Snap Cap
Bottom Shape Conical
Graduations 0.5 to 1.5 mL
Shape Cylindrical
Length (Metric) 3 cm
Product Type Locking Snap-cap Microtube
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.