Learn More
BOLA Micro Reaction Vessels
Click to view available options
For Use With (Equipment):
connections
threads
Unit Size:
Each
Description
Vessel made of translucent, non-porous PFA, screw cap made of PTFE with threaded necks for connection
Specifications
Specifications
| For Use With (Equipment) | threads |
| Autoclavable | Autoclavable |