Learn More
SI Analytics™ Micro-Ostwald Viscometers
Micro-Ostwald viscometers are calibrated for automatic measurement
Brand:  SI Analytics™ 285404322
Additional Details : Weight : 0.10000kg
Product Code. 10102081
  • 360.00€ / Each
Estimated Shipment: 03-06-2024
Description
  • Calibrated for automatic measurement
  • Type No. 517 20
  • Capillary No. II
  • Constant K = 0.1
Specifications
Specifications
Micro-Ostwald Viscometer