Learn More
Cole Parmer MICRO-METERING VALVE WITH FITTINGS, 10-32 CONED, 1/16'' OD TUBING
Brand:  Cole Parmer 02013-90
MICRO-METERING VALVE WITH FITTINGS, 10-32 CONED, 1/16'' OD TUBING
Product Code. 16652712
  • 118.00€ / Pack of 1