Learn More
Epredia™ Micro-Cassette, Coarse Mesh
Product Code. 12543597 Shop All Epredia Products
Click to view available options
Pore Style:
Fine Mesh
Unit Size:
Each
Product Code. 12543597
Brand: Epredia™ 33124
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Easily process your smallest specimens with the Epredia™ Micro-Cassette, constructed entirely of stainless steel.
Highlights:
  • Positive clip-on cover for micro specimen
  • Available with fine or coarse mesh
Dimensions:
  • 0.79 × 0.197″ (20 × 5mm)
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Fine Mesh
Depth (English) 0.197 in.
Depth (Metric) 5 mm
Material Stainless Steel
Diameter (English) 0.79 in.
Diameter (Metric) 20 mm