missing translation for 'onlineSavingsMsg'
Learn More

Mibolerone, TRC

Product Code. 30484006
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484006 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484006 Supplier Toronto Research Chemicals Supplier No. TRCM34200010MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mibolerone
CAS 3704-09-4
Purity Grade Notes HPLC
Molecular Formula C20 H30 O2
Quantity 10 mg
Synonym Estr-4-en-3-one, 17β-hydroxy-7α,17-dimethyl- (7CI,8CI), (7α,17β)-17-Hydroxy-7,17-dimethylestr-4-en-3-one, 7α,17α-Dimethyl-17β-hydroxy-4-estren-3-one, 7α,17α-Dimethyl-19-nortestosterone, Cheque, Matenon, Miboleron, Mibolerone, NSC 72260, U 10997
InChI Formula InChI=1S/C20H30O2/c1-12-10-13-11-14(21)4-5-15(13)16-6-8-19(2)17(18(12)16)7-9-20(19,3)22/h11-12,15-18,22H,4-10H2,1-3H3/t12-,15+,16-,17+,18-,19+,20+/m1/s1
SMILES C[C@@H]1CC2=CC(=O)CC[C@@H]2[C@H]3CC[C@@]4(C)[C@@H](CC[C@]4(C)O)[C@H]13
IUPAC Name (7R,8R,9S,10R,13S,14S,17S)-17-hydroxy-7,13,17-trimethyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 302.45
Formula Weight 302.2246
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.