Learn More
STEMCELL Technologies mFreSR™
Product Code. 15522640
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 5 mL Tubes
50 mL
Unit Size:
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15522640 10 x 5 mL Tubes 50mL
13479008 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15522640 Supplier STEMCELL Technologies Supplier No. 05854
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Defined, serum-free cryopreservation medium for human ES and iPS cells
  • mFreSR™ is a serum-free cryopreservation medium designed for the cryopreservation of human embryonic and induced pluripotent stem cells (ES and iPS cells)
  • mFreSR™ contains DMSO and is complete and ready-to-use
  • Together with mTeSR™1, TeSR™2, or mTeSR™ Plus, mFreSR™ eliminates the use of feeders and serum
  • Human ES and iPS cells cryopreserved in mFreSR™ have thawing efficiencies higher than reported conventional thawing methods using serum
Specifications
Cell Type Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 10 x 5 mL Tubes