Learn More
Mettler Toledo™ Titration Burette For Titration Sensor
Product Code. 10385463 Shop All Mettler Toledo Products
Click to view available options
Volume (Metric):
20 mL
Unit Size:
Each
Product Code. 10385463
Brand: Mettler Toledo™ 51107502
Only null left
Add to basket
Add to basket
Includes
RFID chip
Features optional ceramic valve discs for crystallizing titrants.
  • Allow rapid titrant change
  • Red protection tubing against light
  • Replaceable PTFE valve discs
  • PTFE pistons w/tripple lip seal assure good tightness
Compliance
ISO 8655-3
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Burette
Includes RFID chip
Volume (Metric) 20 mL