Learn More
Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters
Product Code. 13430689 Shop All Mettler Toledo Products
Click to view available options
Type:
Blank Module
Conductivity Module
pH/Ion Module
pH/mV Module
Unit Size:
Each
Product Code. 13430689
Brand: Mettler Toledo™ 30034473
The modular design of SevenExcellence allows you to expand your instrument with additional measurement parameters for future needs at any time.
Just add the new expansion unit, switch on the instrument and the new expansion unit is automatically recognized and ready to use! SevenExcellence fits in total three expansion units in any composition.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy (Temperature) ±0.1°C
Type Conductivity Module
Range (Conductivity) 0.001μS/cm to 2000mS/cm
Range (Salinity) 0 to 80psu
Range (TDS) 0.001mg/L to 1000g/L
Resolution (Conductivity) ±0.5%
Resolution (Temperature) 0.1°C
Temperature (Metric) Range -30°C to +130°C
For Use With (Equipment) SevenExcellence Benchtop Meters