Learn More
Kartell™ Micro Centrifuge Test Tube with Cap
Product Code. 15947870
Click to view available options
Color:
Yellow
Unit Size:
Pack of 10000
Product Code. 15947870
Brand: Kartell™ 0029806
Specifications
Color Yellow
For Use With (Application) Sample analysis
No. per Pack 1000/Pk.
Max. RCF 11.000 x g