Learn More
METTLER TOLEDO™ InLab™ Versatile pH Electrode Replaceable Junction Tool
Product Code. 10396862 Shop All Mettler Toledo Products
Click to view available options
Junction:
Ceramic
Unit Size:
Each
Product Code. 10396862
Brand: METTLER TOLEDO™ 51340267
Tool consisting of two parts to replace ceramic junctions on InLab™ Versatile and InLab™ Versatile Pro.
  • Accessory
  • For use with InLab™ Versatile and InLab™ Versatile Pro
TRUSTED_SUSTAINABILITY
Specifications
Junction Ceramic
Type Replaceable junction tool
For Use With (Application) Alternative for measurements in simple samples, offer an outstanding price/performance ratio
For Use With (Equipment) InLab designations versatile and versatile Pro pH electrode
Temperature (Metric) Range 0°C to 60°C