Learn More
Mettler Toledo™ G20 100mL Plastic Beakers for Titration Stand Kit
Product Code. 11323070 Shop All Mettler Toledo Products
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11323070 100 mL Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11323070 Supplier Mettler Toledo™ Supplier No. 51109388
Plastic beakers for G20 titration stand kit.
For use with G20 titration stand kit
  • 100mL
  • Plastic
TRUSTED_SUSTAINABILITY
Specifications
Type Beaker
Volume (Metric) 100 mL