Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
Product Code. 15927281
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
None
Unit Size:
Each
Product Code. 15927281
Brand: Metro™ TTA18K3
Only null left
Add to basket
Add to basket
Includes
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
High-density, easy access storage
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
Shelves are ordered separately
Specifications
Product Type Stationary Intermediate Unit Kit
Finish Metroseal 3 with Microban™
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Height (English) 86 in.
Height (Metric) 2185 mm
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 7.5 lb.
Weight (Metric) 3.3 kg
Width (English) Shelf 18 in.
Width (Metric) Shelf 45.7 cm
Material Steel