Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Product Code. 15937301
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
Product Code. 15937301
Brand: Metro™ TTM21K3
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
High-density, easy access storage.
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Product Type Mobile Unit Kit
Finish Metroseal 3 with Microban™
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 33 lb.
Weight (Metric) 14.8 kg
Width (English) Shelf 21 in.
Width (English) 23.25 in.
Width (Metric) Shelf 53 cm
Width (Metric) 59.05 cm
Material Steel