Learn More
Metro™ Super Erecta™ Swedged Posts
Product Code. 15946711
Click to view available options
Height (English):
33 in.
54 in.
63 in.
Length (English):
33 in.
54 in.
63 in.
Unit Size:
Each
Product Code. 15946711
Brand: Metro™ 54UPSSW
For cart-washable and autoclave applications.
  • Each Type 304 Stainless Steel post has an aluminum cap swedged into the top
  • For use with standard stem casters
Specifications
Product Type Mobile Shelving Post
For Use With (Application) Autoclavable and Cart Washable Applications
For Use With (Equipment) Metro™ Super Erecta™ stem casters
Height (English) 54 in.
Height (Metric) 137.1 cm
Length (English) 54 in.
Length (Metric) 137 cm
Material Stainless Steel