Learn More
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf
Product Code. 15927031
Click to view available options
Finish:
Epoxy coated
Length (English):
28.3 in.
40.3 in.
Weight (English):
7.5 lb.
10 lb.
Unit Size:
Each
Product Code. 15927031
Brand: Metro™ LT36WE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Expand your workspace in your lab. Metro Stainless Laboratory Worktable shelves come in a choice of stainless steel or epoxy-coated.
  • Shelves mount on three-sided frames with use of clips
  • Load rated for a maximum of 150 lbs.
  • Also compatible with MetroMax i Lab Worktables
Specifications
Product Type Wire Shelf
Finish Epoxy coated
For Use With (Equipment) Metro™ MetroMax i™ and other Stainless Steel worktables
Length (English) 28.3 in.
Length (Metric) 72 cm
Load Bearing Capacity (English) 150 lb.
Load Bearing Capacity (Metric) 68 kg
Mounting Type 3-Sided Frame
Weight (English) 7.5 lb.
Weight (Metric) 3.4 kg