Learn More
Metro™ Stainless Lab Worktable, Stainless Top, With Backsplash and 3-Sided Frame
Product Code. 15917121
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 35 in.
35.75 x 30 x 35 in.
47.75 x 30 x 35 in.
59.75 x 30 x 35 in.
Includes:
Backsplash with 3 sided frame
Backsplash with 3 sided frame, adaptor insert
Backsplash with 3-sided Frame, Adapter Insert
Product Type:
Mobile Table
Stationary Table
Unit Size:
Each
Product Code. 15917121
Brand: Metro™ LTS60US
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure your workers remain safe in the laboratory. Metro Stainless Lab Worktable with backsplash, 3-sided frame and stainless steel construction throughout provides cleanliness along with corrosion-proof performance.
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Stationary Table
Dimensions (L x W x H) 59.75 x 30 x 35 in.
For Use With (Application) Ideal for lab
Height (English) 35 in.
Height (Metric) 89 cm
Includes Backsplash with 3 sided frame
Length (English) 59.75 in.
Length (Metric) 151.7 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel