Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Product Code. 15967331
Click to view available options
Dimensions (L x W x H):
48 x 30 x 34 in.
60 x 30 x 34 in.
60 x 36 x 34 in.
70 x 30 x 34 in.
72 x 30 x 34 in.
96 x 30 x 34 in.
Material:
Stainless Steel
Stainless Steel (14 ga.)
Includes:
Bottom 3-sided frame (SS)
Bottom H-frame (SS)
Bottom shelf (SS)
Unit Size:
Each
Product Code. 15967331
Brand: Metro™ WT366US
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.
  • 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
  • Support structure virtually eliminates drumming
  • Posts with adjustable feet compensate for slight floor irregularities
  • Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
Product Type Stationary Table
Base Style Open with side and rear crossrails
Certifications/Compliance NSF
Dimensions (L x W x H) 60 x 36 x 34 in.
For Use With (Application) Ideal for lab
Height (English) 34 in.
Height (Metric) 86.4 cm
Includes Bottom 3-sided frame (SS)
Length (English) 60 in.
Length (Metric) 91.4 cm
Weight (English) 104 lb.
Weight (Metric) 47 kg
Width (English) 36 in.
Width (Metric) 91.4 cm
Material Stainless Steel