Learn More
Methyltris(trimethylsiloxy)silane, TRC
Specifications
Specifications
| Chemical Name or Material | Methyltris(trimethylsiloxy)silane |
| CAS | 17928-28-8 |
| Purity Grade Notes | GC |
| Molecular Formula | C10H30O3Si4 |
| Quantity | 1 g |
| Synonym | Tris(trimethylsiloxy)methylsilane, 1,1,1,3,5,5,5-Heptamethyl-3-(trimethylsiloxy)trisiloxane, TMF 1.5 |
| InChI Formula | InChI=1S/C10H30O3Si4/c1-14(2,3)11-17(10,12-15(4,5)6)13-16(7,8)9/h1-10H3 |
| SMILES | O([Si](O[Si](C)(C)C)(O[Si](C)(C)C)C)[Si](C)(C)C |
| IUPAC Name | trimethyl-[methyl-bis(trimethylsilyloxy)silyl]oxysilane |
| Molecular Weight (g/mol) | 310.68 |
| Show More |