Learn More
R&D Systems™ Growth Hormone Antibody Pair [HRP]
Product Code. 18611811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611811 5 Plates Pack of 5
18257626 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 18611811 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' NBP2794205Plates
 Restricted Item
Estimated Shipment: 07-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Growth Hormone.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 2.81 pg/mL
Assay Sensitivity 2.81-180 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Line Novus
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Growth Hormone.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Gene ID (Entrez) 2688
Gene Symbol GH1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 24 ng of recombinant Human Growth Hormone.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human Growth Hormone monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human Growth Hormone monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins
    Storage Requirements Storage is content dependent.
    Target Growth Hormone
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 180 pg/mL is recommended.