Learn More
Methylene Violet 3RAX (~90%), TRC
Product Code. 30474977
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474977 500 mg 500mg
30466293 5 g 5g
30476809 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30474977 Supplier Toronto Research Chemicals Supplier No. TRCM236755500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4569-86-2
Molecular Formula C22H23N4 . Cl
Molecular Weight (g/mol) 378.9
Synonym 3-Amino-7-(diethylamino)-5-phenylphenazin-5-ium Chloride, C.I. 50206, Diethylsafranin, 3-Amino-7-diethylamino-5-phenylphenazinium Chloride
IUPAC Name 3-amino-7-(diethylamino)-5-phenylphenazin-5-ium chloride
SMILES CCN(C1=CC2=[N+](C3=CC=CC=C3)C4=CC(N)=CC=C4N=C2C=C1)CC.[Cl-]
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C22H22N4.ClH/c1-3-25(4-2)18-11-13-20-22(15-18)26(17-8-6-5-7-9-17)21-14-16(23)10-12-19(21)24-20;/h5-15,23H,3-4H2,1-2H3;1H
Formula Weight 378.16
Chemical Name or Material Methylene Violet 3RAX