missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadienylmanganese tricarbonyl, 97%

Product Code. 11458630
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11458630

Brand: Thermo Scientific Alfa Aesar 047125.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12108-13-3 | C9H7MnO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12108-13-3
Molecular Formula C9H7MnO3
MDL Number MFCD00001426
Synonym MMT

Specifications

Melting Point 1.5°C
Color Yellow
Density 1.388 g/mL
Boiling Point 233°C
Flash Point 96°C (205°F)
Refractive Index 1.5873
Quantity 5 g
UN Number UN3281
Sensitivity Air sensitive
Merck Index 14,6223
Solubility Information Insoluble in water; Soluble in hydrocarbon solvents, alcohols, ether, acetone
Formula Weight 218.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.