missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tetracosanoate, TRC

Product Code. 30138206
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138206 500 mg 500mg
30133122 1 g 1g
17939344 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138206 Supplier Toronto Research Chemicals Supplier No. TRCM327050500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2442-49-1
Molecular Formula C25 H50 O2
Molecular Weight (g/mol) 382.66
Synonym Methyl lignocerate, Methyl tetracosanoate
IUPAC Name methyl tetracosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C25H50O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25(26)27-2/h3-24H2,1-2H3
Formula Weight 382.3811
Percent Purity >95%
Chemical Name or Material Tetracosanoic acid methyl ester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.